Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferroportin/SLC40A1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP121502Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP121502Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP121502Z Supplier Novus Biologicals Supplier No. NBP121502AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ferroportin/SLC40A1 Polyclonal specifically detects Ferroportin/SLC40A1 in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).

Specifications

Antigen Ferroportin/SLC40A1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular)
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q9NP59
Gene Alias Ferroportin-1, FPN1, FPN1IREG1ferroportin 1, HFE4, HFE4ferroportin-1, IREG1, iron regulated gene 1, Iron-regulated transporter 1, member 3, MST079, MSTP079, MTP1, putative ferroportin 1 variant IIIB, SLC11A3, SLC11A3iron regulated gene 1, solute carrier family 11 (proton-coupled divalent metal ion transporters), solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3, solute carrier family 40 (iron-regulated transporter), member 1, solute carrier family 40 member 1
Gene Symbols SLC40A1
Host Species Rabbit
Immunogen This Ferroportin/SLC40A1 Antibody was developed against a synthetic peptide made to an internal portion of human Ferroportin 1 (within residues 250-300). [Swiss-Prot: Q9NP59]
Molecular Weight of Antigen 62 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 30061
Target Species Human, Mouse, Pig, Bovine
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.