Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Ferrostatin 1

Click to view available options
Quantity:
10 mg
50 mg
Description
Ferrostatin 1 is a selective inhibitor of erastin induced ferroptosis (EC50 = 60 nM). Specifically inhibits Ras selective lethal compound -induced death, but not cell death induced by other oxidative lethal compounds and apoptosis-inducing agents. Inhibits ferroptosis in cancer cells; also inhibits glutamate-induced cell death in organotypic rat brain slices. Prevents erastin induced accumulation of cytosolic and lipid reactive oxygen species.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | Ferrostatin 1 |
| Purity | >98% |
| Molecular Weight (g/mol) | 262.35 |
| Inhibitors | Ferrostatin 1 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction