Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ferrostatin 1
GREENER_CHOICE

Catalog No. 518010
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 51-801-0

Supplier: Tocris Bioscience™ 5180/10

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective inhibitor of erastin induced ferroptosis

Ferrostatin 1 is a selective inhibitor of erastin induced ferroptosis (EC50 = 60 nM). Specifically inhibits Ras selective lethal compound -induced death, but not cell death induced by other oxidative lethal compounds and apoptosis-inducing agents. Inhibits ferroptosis in cancer cells; also inhibits glutamate-induced cell death in organotypic rat brain slices. Prevents erastin induced accumulation of cytosolic and lipid reactive oxygen species.

Specifications

Quantity 10 mg
Product Type Ferrostatin 1
Purity >98%
Molecular Weight (g/mol) 262.35
Inhibitors Ferrostatin 1

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.