Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Ammonium Sulfate Titrant, For Chlorine, Chlorine Dioxide (1mL = 0.1mg Cl), Certified, 0.00282N ±0.00010N (0.00282M), LabChem™

$66.70 - $189.40

Specifications

Color Clear pale green
Concentration 0.00282N ±0.00010N (0.00282M)
Format Liquid
Packaging Amber Glass
Storage Requirements Room Temperature
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
LC145501
View Documents
LabChem, Inc.
LC145501
500 mL
Each for $66.70
Only null left
Add to Cart
 
LC145502
View Documents
LabChem, Inc.
LC145502
1 L
Each for $85.10
Only null left
Add to Cart
 
LC145504
View Documents
LabChem, Inc.
LC145504
4 L
Each for $189.40
Only null left
Add to Cart
 
Description

Description

Specifications

Specifications

Clear pale green
Liquid
Room Temperature
0.00282N ±0.00010N (0.00282M)
Amber Glass
Ferrous Ammonium Sulfate Titrant
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.