Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fes Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB086469 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB086469 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB086469 Supplier Novus Biologicals Supplier No. NBP299957JF646

Rabbit Polyclonal Antibody

Fes Polyclonal specifically detects Fes in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Fes
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50 mM Sodium Borate
Gene Alias EC 2.7.10, EC 2.7.10.2, feline sarcoma oncogene, feline sarcoma virus, Proto-oncogene c-Fes, Proto-oncogene c-Fps, proto-oncogene tyrosine-protein kinase Fes/Fps, tyrosine-protein kinase Fes/Fps
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Fes (P07332-1; Met1-Arg822).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2242
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.