Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fetuin A/AHSG Antibody (2), DyLight 488
SDP

Catalog No. NB073231 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB073231 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB073231 Supplier Novus Biologicals Supplier No. NBP290414G

Rabbit Monoclonal Antibody

Fetuin A/AHSG Monoclonal specifically detects Fetuin A/AHSG in Mouse samples. It is validated for ELISA.

Specifications

Antigen Fetuin A/AHSG
Applications ELISA
Classification Monoclonal
Clone 2
Conjugate DyLight 488
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias A2HS, AHS, alpha-2-HS-glycoprotein, alpha-2-Z-globulin, Ba-alpha-2-glycoprotein, FETUAba-alpha-2-glycoprotein, fetuin-A, HSGA
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Fetuin A/AHSG (NP_038493.1; Met 1-Ile 345) .
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 197
Test Specificity No cross-reactivity in ELISA with: Human Fetuin-A/ AHSG
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.