Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fetuin A/AHSG Antibody, Novus Biologicals™
SDP

Catalog No. NBP10215201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP10215201 0.125 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP10215201 Supplier Novus Biologicals Supplier No. NBP1021520.125MG
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Fetuin A/AHSG Polyclonal specifically detects Fetuin A/AHSG in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Fetuin A/AHSG
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:10000-1:20000, ELISA 1:40.000-1:200000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide
Gene Alias A2HS, AHS, alpha-2-HS-glycoprotein, alpha-2-Z-globulin, Ba-alpha-2-glycoprotein, FETUAba-alpha-2-glycoprotein, fetuin-A, HSGA
Gene Symbols AHSG
Host Species Goat
Immunogen This purified antibody was prepared from rabbit serum after repeated immunizations with a recombinant human fetuin (a2-HS glycoprotein) processed to remove a 40 amino acid residue bridging peptide resulting in the mature form of the protein.
Quantity 0.125 mg
Primary or Secondary Primary
Gene ID (Entrez) 197
Test Specificity Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Goat Serum.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.