Learn More
Description
Specifications
Specifications
| Antigen | FFAR1/GPR40 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | free fatty acid receptor 1, G protein-coupled receptor 40, GPCR40, GPR40FFA1R, G-protein coupled receptor 40 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human FFAR1/GPR40 (NP_005294.1). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
