Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FFAR1/GPR40 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$207.00 - $499.50
Specifications
Antigen | FFAR1/GPR40 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
FFAR1/GPR40 Polyclonal specifically detects FFAR1/GPR40 in Human samples. It is validated for Western Blot.Specifications
FFAR1/GPR40 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
free fatty acid receptor 1, G protein-coupled receptor 40, GPCR40, GPR40FFA1R, G-protein coupled receptor 40 | |
A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human FFAR1/GPR40 (NP_005294.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
GPCR | |
PBS with 50% glycerol, pH7.3. | |
2864 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title