Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FFAR2/GPR43 Antibody (1000624), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB340201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340201 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340201 Supplier Novus Biologicals Supplier No. FAB10082MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FFAR2/GPR43 Monoclonal antibody specifically detects FFAR2/GPR43 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen FFAR2/GPR43
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 1000624
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias free fatty acid activated receptor 2, free fatty acid receptor 2, G protein-coupled receptor 43, GPR43FFA2R, G-protein coupled receptor 43
Host Species Mouse
Immunogen NS0 Mouse myeloma cell line transfected with human FFAR2/GPR43, Met1-Glu330, Accession # O15552
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2867
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.