Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FFAR4/GPR120 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NP10858A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP10858A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP10858A405 Supplier Novus Biologicals Supplier No. NBP100858AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FFAR4/GPR120 Polyclonal specifically detects FFAR4/GPR120 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.

Specifications

Antigen FFAR4/GPR120
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias G protein-coupled receptor 120, G protein-coupled receptor 129, G protein-coupled receptor PGR4, GPR120, GPR129, G-protein coupled receptor 120, G-protein coupled receptor 129, G-protein coupled receptor GT01, G-protein coupled receptor PGR4, G-protein-coupled receptor GT01, GT01, MGC119984, omega-3 fatty acid receptor 1, PGR4DKFZp686F0824
Gene Symbols FFAR4
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human GPR120 protein (between residues 200-300) [UniProt Q5NUL3]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 338557
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.