Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FFAR4/GPR120 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB166283 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166283 100 μg
NB166284 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166283 Supplier Novus Biologicals Supplier No. NBP315594100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FFAR4/GPR120 Polyclonal antibody specifically detects FFAR4/GPR120 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen FFAR4/GPR120
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias G protein-coupled receptor 120, G protein-coupled receptor 129, G protein-coupled receptor PGR4, GPR120, GPR129, G-protein coupled receptor 120, G-protein coupled receptor 129, G-protein coupled receptor GT01, G-protein coupled receptor PGR4, G-protein-coupled receptor GT01, GT01, MGC119984, omega-3 fatty acid receptor 1, PGR4DKFZp686F0824
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence within amino acids 298-377 of human FFAR4/GPR120 (NP_859529.2).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 338557
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.