Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FFAR4/GPR120 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$159.60 - $389.00
Specifications
Antigen | FFAR4/GPR120 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
FFAR4/GPR120 Polyclonal antibody specifically detects FFAR4/GPR120 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
FFAR4/GPR120 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
G protein-coupled receptor 120, G protein-coupled receptor 129, G protein-coupled receptor PGR4, GPR120, GPR129, G-protein coupled receptor 120, G-protein coupled receptor 129, G-protein coupled receptor GT01, G-protein coupled receptor PGR4, G-protein-coupled receptor GT01, GT01, MGC119984, omega-3 fatty acid receptor 1, PGR4DKFZp686F0824 | |
Recombinant fusion protein containing a sequence within amino acids 298-377 of human FFAR4/GPR120 (NP_859529.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
GPCR | |
PBS, 50% glycerol, pH7.3 | |
338557 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title