Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF-13 Antibody (OTI7E8), PE, Novus Biologicals™
SDP

Catalog No. NB289413 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289413 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289413 Supplier Novus Biologicals Supplier No. NBP270721PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGF-13 Monoclonal antibody specifically detects FGF-13 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen FGF-13
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI7E8
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13
Host Species Mouse
Immunogen Full length human recombinant protein of human FGF13(NP_004105) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2258
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.