Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF-19 Antibody (032), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB069601 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB069601 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB069601 Supplier Novus Biologicals Supplier No. NBP290078AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

FGF-19 Monoclonal specifically detects FGF-19 in Human samples. It is validated for ELISA.

Specifications

Antigen FGF-19
Applications ELISA
Classification Monoclonal
Clone 32
Conjugate Alexa Fluor 488
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias FGF-19, fibroblast growth factor 19
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human FGF-19 (O95750; Phe27-Lys216).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9965
Test Specificity No cross-reactivity in ELISA with: Human FGF6 Human FGF9 Human FGF10 Human FGF17 Human FGF18 Human FGF acidic Human FGF basic
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.