Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-19 Antibody (MM0285-4G16) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2122990.025MG
Description
FGF-19 Monoclonal specifically detects FGF-19 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FGF-19 | |
Monoclonal | |
LYOPH | |
Western Blot 1:500-1000, Immunohistochemistry 1:20-100, Immunohistochemistry-Paraffin 1:20-100 | |
O95750 | |
FGF19 | |
Human FGF19 | |
0.025 mg | |
Angiogenesis, Neuroscience | |
9965 | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
MM0285-4G16 | |
Unconjugated | |
A 0.2 um filtered solution in PBS with No Preservative | |
FGF-19, fibroblast growth factor 19 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction