Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-19 Antibody (MM0285-4G16) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
Antigen | FGF-19 |
---|---|
Clone | MM0285-4G16 |
Concentration | LYOPH |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Description
FGF-19 Monoclonal specifically detects FGF-19 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FGF-19 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Human | |
FGF-19, fibroblast growth factor 19 | |
FGF19 | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
MM0285-4G16 | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Angiogenesis, Neuroscience | |
O95750 | |
9965 | |
IgG1 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title