Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF-21 Antibody (005), Alexa Fluor™ 488
SDP

Catalog No. NB075204 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB075204 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB075204 Supplier Novus Biologicals Supplier No. NBP290579AF488

Rabbit Monoclonal Antibody

FGF-21 Monoclonal specifically detects FGF-21 in Mouse samples. It is validated for ELISA.

Specifications

Antigen FGF-21
Applications ELISA
Classification Monoclonal
Clone 5
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias FGF-21, fibroblast growth factor 21
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse FGF-21 (Q9JJN1; Met1-Ser210).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 26291
Test Specificity Has cross-reactivity in ELISA with: Human FGF21
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.