Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF-21 Antibody (461804), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB364853 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364853 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364853 Supplier Novus Biologicals Supplier No. FAB25372D
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGF-21 Monoclonal specifically detects FGF-21 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry.

Specifications

Antigen FGF-21
Applications CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry
Classification Monoclonal
Clone 461804
Conjugate DyLight 350
Dilution Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FGF-21, fibroblast growth factor 21
Host Species Mouse
Immunogen E. coli-derived recombinant human FGF-21, His29-Ser209, Accession # Q9NSA1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 26291
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.