Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF-22 Antibody (MM0286-4F28), PE, Novus Biologicals™
SDP

Catalog No. NB361063 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361063 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361063 Supplier Novus Biologicals Supplier No. NBP212300PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGF-22 Monoclonal antibody specifically detects FGF-22 in Human samples. It is validated for Western Blot, Neutralization

Specifications

Antigen FGF-22
Applications Western Blot, Neutralization
Classification Monoclonal
Clone MM0286-4F28
Conjugate PE
Dilution Western Blot, Block/Neutralize
Formulation PBS
Gene Alias FGF-22, fibroblast growth factor 22
Host Species Mouse
Immunogen Human recombinant FGF-22
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 27006
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.