Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF-23 Antibody (FGF23/4168), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB380947 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB380947 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB380947 Supplier Novus Biologicals Supplier No. NBP308383AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGF-23 Monoclonal antibody specifically detects FGF-23 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen FGF-23
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone FGF23/4168
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias ADHR, FGF-23, fibroblast growth factor 23, HPDR2, HYPF, Phosphatonin, PHPTC, tumor-derived hypophosphatemia inducing factor, Tumor-derived hypophosphatemia-inducing factor
Host Species Mouse
Immunogen Recombinant fragment (around aa25-251) of human FGF-23 protein (exact sequence is proprietary) (Uniprot: Q9GZV9)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8074
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.