Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF-3 Antibody (MSD1), HRP, Novus Biologicals™
SDP

Catalog No. NB200603H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200603H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB200603H Supplier Novus Biologicals Supplier No. NB200603H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGF-3 Monoclonal specifically detects FGF-3 in Human, Mouse, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen FGF-3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MSD1
Conjugate HRP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with No Preservative
Gene Alias fibroblast growth factor 3, fibroblast growth factor 3 (murine mammary tumor virus integration site (v-int-2) oncogene homolog), HBGF-3, mouse, oncogene INT2
Gene Symbols FGF3
Host Species Mouse
Immunogen Peptide conjugated to BSA.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2248
Test Specificity This antibody recognizes all naturally occurring FGF-3 variants and all recombinant FGF-3 proteins.
Target Species Human, Mouse, Xenopus species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.