Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF-4 Antibody (3A12G9) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP237374

 View more versions of this product

Catalog No. NBP237374


Only null left
Add to Cart

Description

Description

FGF-4 Monoclonal specifically detects FGF-4 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

FGF-4
Monoclonal
1 mg/mL
Western Blot 1:500 - 1:2000, ELISA 1:10000
fibroblast growth factor 4, HBGF-4Transforming protein KS3, heparin secretory transforming protein 1, Heparin secretory-transforming protein 1, Heparin-binding growth factor 4, HST-1HSTF-1, HSTF1fibroblast growth factor 4 splice isoform, HSTFGF-4, human stomach cancer, transforming factor from FGF-related oncogene, kaposi sarcoma oncogene, KFGF, K-FGF, KS3, oncogene HST
Mouse
22 kDa
0.1 mL
Angiogenesis, Biologically Active Proteins, Cancer
2249
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, ELISA
3A12G9
Unconjugated
PBS with 0.05% Sodium Azide
FGF4
Purified recombinant fragment of humanFGF4 (AA: 62-123) expressed in E. Coli.
Affinity Purified
RUO
Primary
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only