Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-4 Antibody (3A12G9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | FGF-4 |
---|---|
Clone | 3A12G9 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
Applications | Western Blot, ELISA |
Description
FGF-4 Monoclonal specifically detects FGF-4 in Human samples. It is validated for Western Blot, ELISA.Specifications
FGF-4 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
Human | |
fibroblast growth factor 4, HBGF-4Transforming protein KS3, heparin secretory transforming protein 1, Heparin secretory-transforming protein 1, Heparin-binding growth factor 4, HST-1HSTF-1, HSTF1fibroblast growth factor 4 splice isoform, HSTFGF-4, human stomach cancer, transforming factor from FGF-related oncogene, kaposi sarcoma oncogene, KFGF, K-FGF, KS3, oncogene HST | |
FGF4 | |
IgG1 | |
Affinity Purified |
3A12G9 | |
Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
Monoclonal | |
Mouse | |
Angiogenesis, Biologically Active Proteins, Cancer | |
PBS with 0.05% Sodium Azide | |
2249 | |
Purified recombinant fragment of humanFGF4 (AA: 62-123) expressed in E. Coli. | |
Primary | |
22 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title