Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP198443
Description
FGF-8 Polyclonal specifically detects FGF-8 in Human, Mouse samples. It is validated for Western Blot.Specifications
| FGF-8 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| AIGFKAL6, Androgen-induced growth factor, FGF-8, fibroblast growth factor 8, fibroblast growth factor 8 (androgen-induced), HBGF-8, Heparin-binding growth factor 8, MGC149376 | |
| Rabbit | |
| 24 kDa | |
| 100 μL | |
| Angiogenesis, Cell Cycle and Replication | |
| 2253 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_149354 | |
| FGF8 | |
| The immunogen for this antibody is FGF8 - middle region. Peptide sequence LIVETDTFGSRVRVRGAETGLYICMNKKGKLIAKSNGKGKDCVFTEIVLE. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Zebrafish: 79%. | |
| Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction