Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF-9 Antibody (FG9-77), Biotin, Novus Biologicals™
SDP

Catalog No. NB600623B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600623B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600623B Supplier Novus Biologicals Supplier No. NB600623B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGF-9 Monoclonal specifically detects FGF-9 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen FGF-9
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone FG9-77
Conjugate Biotin
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias FGF-9, Fibroblast growth factor 9, fibroblast growth factor 9 (glia-activating factor), GAF, glia-activating factor, HBFG-9, HBGF-9, Heparin-binding growth factor 9, MGC119914, MGC119915, SYNS3
Gene Symbols FGF9
Host Species Mouse
Immunogen Recombinant mouse FGF-9
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 2254
Test Specificity Monoclonal Anti-FGF9 reacts specifically with recombinant FGF9 (fibroblast growth factor 9). It does not cross-react with human FGF Acidic and FGF Basic.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.