Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF acidic/FGF1 Antibody (OTI4D2), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP270717A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP270717A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP270717A94 Supplier Novus Biologicals Supplier No. NBP270717AF594

Mouse Monoclonal Antibody

FGF acidic/FGF1 Monoclonal specifically detects FGF acidic/FGF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FGF acidic/FGF1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4D2
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias AFGF, alpha, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1
Gene Symbols FGF1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 16-155 of human FGF acidic/FGF1 (NP_000791) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2246
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.