Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF acidic/FGF1 Antibody (V8P1G5*F2), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP20406A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20406A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20406A30 Supplier Novus Biologicals Supplier No. NBP250406AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGF acidic/FGF1 Monoclonal specifically detects FGF acidic/FGF1 in Zebrafish samples. It is validated for ELISA, Immunohistochemistry.

Specifications

Antigen FGF acidic/FGF1
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone V8P1G5*F2
Conjugate Alexa Fluor 350
Dilution ELISA, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias AFGF, alpha, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1
Gene Symbols FGF1
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide LPRQVEQEED (C-terminal sequence)
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2246
Target Species Zebrafish
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.