Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF acidic/FGF1 Antibody (V8P1G5*F2), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309457 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309457 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309457 Supplier Novus Biologicals Supplier No. NBP250406MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGF acidic/FGF1 Monoclonal antibody specifically detects FGF acidic/FGF1 in Zebrafish samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen FGF acidic/FGF1
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone V8P1G5*F2
Conjugate mFluor Violet 500 SE
Dilution ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias AFGF, alpha, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide LPRQVEQEED (C-terminal sequence)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2246
Target Species Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.