Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF basic/FGF2/bFGF Antibody (013), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB308448 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308448 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308448 Supplier Novus Biologicals Supplier No. NBP289192MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

FGF basic/FGF2/bFGF Monoclonal antibody specifically detects FGF basic/FGF2/bFGF in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen FGF basic/FGF2/bFGF
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 013
Conjugate mFluor Violet 450 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, synthetic peptide corresponding to the N-terminus of the Human FGF basic/FGF2/bFGF.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2247
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.