Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF basic/FGF2/bFGF Antibody (FB-8), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB6001128G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001128G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001128G Supplier Novus Biologicals Supplier No. NB6001128G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGF basic/FGF2/bFGF Monoclonal specifically detects FGF basic/FGF2/bFGF in Human, Bovine samples. It is validated for Western Blot, ELISA.

Specifications

Antigen FGF basic/FGF2
Applications Western Blot, ELISA
Classification Monoclonal
Clone FB-8
Conjugate DyLight 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2
Gene Symbols FGF2
Host Species Mouse
Immunogen Recombinant full length protein (Human).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2247
Test Specificity Does not cross-react with human fibroblast growth factor - acidic (aFGF).
Target Species Human, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.