Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF basic/FGF2/bFGF Antibody (OTI3D9), HRP, Novus Biologicals™
SDP

Catalog No. NBP270718H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270718H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270718H Supplier Novus Biologicals Supplier No. NBP270718H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGF basic/FGF2/bFGF Monoclonal specifically detects FGF basic/FGF2/bFGF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen FGF basic/FGF2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3D9
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2
Gene Symbols FGF2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 10-155 of human FGF basic/FGF2 (NP_001997) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2247.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.