Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF basic/FGF2/bFGF Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP1978370.025MG
Description
FGF basic/FGF2/bFGF Polyclonal specifically detects FGF basic/FGF2/bFGF in Mouse, Rat samples. It is validated for Western Blot.Specifications
FGF basic/FGF2 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2), 0.5% BSA, 5 mM EDTA, 30% Glycerol with 0.01% Thimerosal | |
Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
2247 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/mL | |
Western Blot 0.5 - 4 ug/mL | |
P13109 | |
FGF2 | |
E. coli-expressed recombinant rat FGF-2 | |
0.025 mg | |
Primary | |
Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction