Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF basic/FGF2/bFGF Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$182.00 - $464.00
Specifications
Antigen | FGF basic/FGF2 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5 - 4 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
FGF basic/FGF2/bFGF Polyclonal specifically detects FGF basic/FGF2/bFGF in Mouse, Rat samples. It is validated for Western Blot.Specifications
FGF basic/FGF2 | |
Western Blot 0.5 - 4 ug/mL | |
Polyclonal | |
Rabbit | |
Mouse, Rat | |
P13109 | |
2247 | |
E. coli-expressed recombinant rat FGF-2 | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
PBS (pH 7.2), 0.5% BSA, 5 mM EDTA, 30% Glycerol with 0.01% Thimerosal | |
Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2 | |
FGF2 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title