Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF-BP Antibody (OTI2B6), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N271527J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271527J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271527J549 Supplier Novus Biologicals Supplier No. NBP271527JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGF-BP Monoclonal specifically detects FGF-BP in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen FGF-BP
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2B6
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 17 kDa heparin-binding growth factor-binding protein, FGF-BP, FGF-BP1, FGFBP-1, FGFBPFGF-binding protein 1, fibroblast growth factor binding protein 1, fibroblast growth factor-binding protein 1, HBp17, HBP1717 kDa HBGF-binding protein, heparin-binding growth factor binding protein
Gene Symbols FGFBP1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 24-234 of human FGF-BP (NP_005121) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9982.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.