Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF14 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB091450 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB091450 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB091450 Supplier Novus Biologicals Supplier No. NBP300172AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FGF14 Polyclonal specifically detects FGF14 in Human samples. It is validated for ELISA.

Specifications

Antigen FGF14
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias bA397O8.2, FGF-14, FHF4FHF-4, fibroblast growth factor 14, Fibroblast growth factor homologous factor 4, SCA27MGC119129
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant FGF14 (NP_787125.1; Lys64-Thr252).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2259
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.