Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | FGF14 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP16900620
![]() |
Novus Biologicals
NBP16900620UL |
20 μL |
Each for $206.00
|
|
|||||
NBP169006
![]() |
Novus Biologicals
NBP169006 |
100 μL |
Each for $487.50
|
|
|||||
Description
FGF14 Polyclonal specifically detects FGF14 in Human samples. It is validated for Western Blot.Specifications
FGF14 | |
Polyclonal | |
Rabbit | |
Angiogenesis, Neuroscience | |
bA397O8.2, FGF-14, FHF4FHF-4, fibroblast growth factor 14, Fibroblast growth factor homologous factor 4, SCA27MGC119129 | |
FGF14 | |
IgG | |
28 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q92915 | |
2259 | |
Synthetic peptides corresponding to FGF14 (fibroblast growth factor 14) The peptide sequence was selected from the middle region of FGF14. Peptide sequence ENYYVIYSSMLYRQQESGRAWFLGLNKEGQAMKGNRVKKTKPAAHFLPKP. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title