Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech FGF16 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. 502718654
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-8654 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-271-8654 Supplier PeproTech Supplier No. 500P160GBT500UG
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

AA Sequence of recombinant protein: AEVGGVFASL DWDLHGFSSS LGNVPLADSP GFLNERLGQI EGKLQRGSPT DFAHLKGILR RRQLYCRTGF HLEIFPNGTV HGTRHDHSRF GILEFISLAV GLISIRGVDS GLYLGMNERG ELYGSKKLTR ECVFREQFEE NWYNTYASTL YKHSDSERQY YVALNKDGSP REGYRTKRHQ KFTHFLPRPV DPSKLPSMSR DLFHYR. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human FGF-16. Anti-Human FGF-16-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human FGF-16 by sandwich ELISA (using 100 μL/well) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human FGF-16 (500-P160G) as a capture antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human FGF-16 Western Blot: To detect hFGF-16 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hFGF-16 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P160GBT-1MG will be provided as 2 x 500 μg

FGF-16 is a heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-16 signals through FGFR 2c and 3c. FGF-16 plays a role in the development of the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF16
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene FGF16
Gene Accession No. O43320
Gene Alias FGF; FGF16; FGF-16; Fibroblast growth factor; Fibroblast growth factor 16; metacarpal 4-5 fusion; MF4
Gene Symbols FGF16
Host Species Goat
Immunogen E.coli-derived Recombinant Human FGF-16.
Purification Method Antigen affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8823
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.