Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF18 Rabbit anti-Mouse, Alexa Fluor 700, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB052912 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB052912 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB052912 Supplier Novus Biologicals Supplier No. NBP298225AF700

Rabbit Polyclonal Antibody

FGF18 Polyclonal specifically detects FGF18 in Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen FGF18
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias FGF-18ZFGF5, fibroblast growth factor 18, zFGF5
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant FGF18 (NP_032031.1; Met1-Gly207).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8817
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.