Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFBP2 Antibody, Novus Biologicals™
SDP

Catalog No. NB419048 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB419048 25ul
NBP181341 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB419048 Supplier Novus Biologicals Supplier No. NBP18134125UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FGFBP2 Polyclonal specifically detects FGFBP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FGFBP2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias 37 kDa killer-specific secretory protein, FGF-binding protein 2, FGF-BP2, FGFBP-2, fibroblast growth factor binding protein 2, fibroblast growth factor-binding protein 2, HBp17-RP, HBP17RP, killer-specific secretory protein of 37 kDa, Ksp37, KSP37HBp17-related protein
Gene Symbols FGFBP2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:PVLRPSVCREAGPQAHMQQVTSSLKGSPEPNQQPEAGTPSLRPKATVKLTEATQLGKDSMEELGKAKPTTR
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 83888
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.