Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR1 alpha Antibody (1058809), Biotin, Novus Biologicals™
SDP

Catalog No. NB297385 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297385 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297385 Supplier Novus Biologicals Supplier No. FAB11336B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGFR1 alpha Monoclonal specifically detects FGFR1 alpha in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen FGFR1 alpha
Applications Flow Cytometry
Classification Monoclonal
Clone 1058809
Conjugate Biotin
Dilution Flow Cytometry
Formulation PBS
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived human FGFR1 alpha, Arg22-Glu376, Accession # P11362.3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2260
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.