Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR1 Antibody (M17D10), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N102080A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N102080A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N102080A350 Supplier Novus Biologicals Supplier No. NB1002080AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGFR1 Monoclonal specifically detects FGFR1 in Human, Mouse samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen FGFR1
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone M17D10
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias basic fibroblast growth factor receptor 1, BFGFR, bFGF-R-1, CD331, CD331 antigen, CEK, EC 2.7.10, EC 2.7.10.1, FGFBR, FGFR-1, fibroblast growth factor receptor 1, FLGH3, FLJ99988, FLT-2, FLT2H4, Fms-like tyrosine kinase 2, fms-related tyrosine kinase 2, H2, H5, HBGFR, heparin-binding growth factor receptor, hydroxyaryl-protein kinase, KAL 2, KAL2, N-SAM, OGD, Proto-oncogene c-Fgr, soluble FGFR1 variant 1, soluble FGFR1 variant 2
Gene Symbols FGFR1
Host Species Mouse
Immunogen Recombinant human ectodomain of FGFr1b expressed in E. coli from pro23 to his415; antigen contained NH2-terminal gly-ser-pro-gly-ile and COOH-terminal glu-phe sequences.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2260
Test Specificity Reacts with both alpha and beta isoforms.
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.