Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR1OP Antibody (SR1871), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32256850UL
Description
FGFR1OP Monoclonal antibody specifically detects FGFR1OP in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
FGFR1OP | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence | |
SR1871 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
FGFR1 oncogene partner, FOPfibroblast growth factor receptor 1 oncogene partner | |
A synthesized peptide derived from human FGFR1OP (Uniprot #: O95684) | |
50 μg | |
Angiogenesis, Apoptosis, Cell Cycle and Replication | |
11116 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction