Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR1OP Antibody (SR1871), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | FGFR1OP |
---|---|
Clone | SR1871 |
Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Description
FGFR1OP Monoclonal antibody specifically detects FGFR1OP in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
FGFR1OP | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
FGFR1 oncogene partner, FOPfibroblast growth factor receptor 1 oncogene partner | |
A synthesized peptide derived from human FGFR1OP (Uniprot #: O95684) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1871 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Angiogenesis, Apoptosis, Cell Cycle and Replication | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
11116 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title