Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR2 alpha Antibody (1057960), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB305327 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305327 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305327 Supplier Novus Biologicals Supplier No. FAB111192Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGFR2 alpha Monoclonal specifically detects FGFR2 alpha in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen FGFR2 alpha
Applications Flow Cytometry
Classification Monoclonal
Clone 1057960
Conjugate DyLight 755
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias BBDS,BEK,BFR-1,CD332,CEK3,CFD1,ECT1,Fibroblast growth factor receptor,Fibroblast growth factor receptor 2,JWS,KGFR,K-SAM,Receptor protein-tyrosine kinase,TK14,TK25
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human FGFR2 alpha, Arg22-Glu377, Accession # P21802.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2263
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.