Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR2 Antibody (7C24), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBT94154UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBT94154UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBT94154UV Supplier Novus Biologicals Supplier No. NB11094154UV
Add to Cart
Add to Cart

Rat Monoclonal Antibody

FGFR2 Monoclonal specifically detects FGFR2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FGFR2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7C24
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P21803
Gene Alias BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25
Gene Symbols FGFR2
Host Species Rat
Immunogen Mouse FGFR2 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2263
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.