Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR2 Antibody (OTI5C5), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N270731A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N270731A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N270731A647 Supplier Novus Biologicals Supplier No. NBP270731AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGFR2 Monoclonal specifically detects FGFR2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FGFR2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI5C5
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25
Gene Symbols FGFR2
Host Species Mouse
Immunogen Full length recombinant protein of human FGF R2 (NP_000132) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2263.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.