Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR2 Rabbit anti-Human, Mouse, Rat, Clone: JM10-60, Novus Biologicals™
SDP

Catalog No. NBP266838 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266838 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266838 Supplier Novus Biologicals Supplier No. NBP266838

Rabbit Monoclonal Antibody

FGFR2 Monoclonal specifically detects FGFR2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen FGFR2
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone JM10-60
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoprecipitation 1:10 - 1:500
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25
Gene Symbols FGFR2
Host Species Rabbit
Immunogen Synthetic peptide within Human FGFR2 aa 30-70/ 821. (SwissProt: P21802 Human; SwissProt: P21803 Mouse; Entrez Gene: 25022 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Angiogenesis, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2263
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.