Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR3 Antibody (OTI1B10), PE, Novus Biologicals™
SDP

Catalog No. NB289410 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289410 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289410 Supplier Novus Biologicals Supplier No. NBP270722PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGFR3 Monoclonal antibody specifically detects FGFR3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen FGFR3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone OTI1B10
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 23-375 of human FGFR3 (NP_000133) produced in SF9 cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2261
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.