Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR4 Antibody (249), PerCP, Novus Biologicals™
SDP

Catalog No. NB307085 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB307085 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB307085 Supplier Novus Biologicals Supplier No. NBP289557PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

FGFR4 Monoclonal antibody specifically detects FGFR4 in Human samples. It is validated for ELISA

Specifications

Antigen FGFR4
Applications ELISA
Classification Monoclonal
Clone 249
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias CD334, CD334 antigen, EC 2.7.10, EC 2.7.10.1, FGFR-4, fibroblast growth factor receptor 4, JTK2hydroxyaryl-protein kinase, MGC20292, protein-tyrosine kinase, TKF, tyrosine kinase related to fibroblast growth factor receptor, tyrosylprotein kinase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human FGFR4 (Accession#: NP_002002.3; Met1-Asp369).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2264
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.